Learn More
Medchemexpress LLC HY-100853 5mg Medchemexpress, IWP-O1 CAS:2074607-48-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1008535MG
Encompass_Preferred
Medchemexpress, HY-100853 5mg IWP-O1 CAS:2074607-48-8 IWP-O1 is a highly potent Porcupine (Porcn) inhibitor, with an EC 50 of 80 pM in L-Wnt-STF cells, suppressing the phosphorylation of Dvl2/3 and LRP6 in HeLa cells. IWP-O1 functions by preventing the secretion of Wnt proteins [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1972