Learn More
TARGETMOL CHEMICALS INC IWR-1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T265125MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IWR-1 (IWR-1-endo) a Wnt pathway inhibitor stabilizes the Axin destruction complex(EC50=0.2 uM). purity: 99%
Catalog No. 50-258-0693