Learn More
Selleck Chemical LLC IWR-1-endo 10mg 1127442-82-3
SDP
Supplier:  Selleck Chemical LLC S708610MG
Encompass_Preferred
IWR-1-endo is a Wnt pathway inhibitor with IC50 of 180 nM in L-cells expressing Wnt3A, induces Axin2 protein levels and promotes ?-catenin phosphorylation by stabilizing Axin-scaffolded destruction complexes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1319406