Learn More
Apexbio Technology LLC IWR-1-endo, 1127442-82-3, MFCD18086875, 10 mg
SDP
Catalog No. 501014191
Encompass_Preferred
MF: C25H19N3O3, MW: 409.44, Solubility: 20.45mg/mL in DMSO. IWR-1-endo is a small molecule inhibitor of Wnt Response with IC50 value of 180nM.
Catalog No. 50-101-4191 Supplier Apexbio Technology LLC Supplier No. B230610
MF: C25H19N3O3, MW: 409.44, Solubility: 20.45mg/mL in DMSO. IWR-1-endo is a small molecule inhibitor of Wnt Response with IC50 value of 180nM.