Learn More
AdipoGen IWR-1-endo
SDP
Catalog No. 502562274
Encompass_Preferred
Chemical. CAS 1127442-82-3. Formula C25H19N3O3. MW 409.4. Potent and reversible cell permeable Wnt pathway signaling inhibitor. Inhibits Wnt-induced accumulation of beta-catenin, leading to proteasomal degradation of this protein through a destruction complex which consists of Apc, Axin2, CK1 and GSK-3beta. Stabilizes the destruction complex, increasing the level of Axin2 protein without changing the levels of Apc or GSK-3beta. Tankyrase-1 TNKS1/PARP5a and Tankyrase-2 TNKS2/PARP5b inhibitor in vitro auto-PARsylation assay.
Catalog No. 50-256-2274 Supplier AdipoGen Supplier No. AGCR13581M005
Chemical. CAS 1127442-82-3. Formula C25H19N3O3. MW 409.4. Potent and reversible cell permeable Wnt pathway signaling inhibitor. Inhibits Wnt-induced accumulation of beta-catenin, leading to proteasomal degradation of this protein through a destruction complex which consists of Apc, Axin2, CK1 and GSK-3beta. Stabilizes the destruction complex, increasing the level of Axin2 protein without changing the levels of Apc or GSK-3beta. Tankyrase-1 TNKS1/PARP5a and Tankyrase-2 TNKS2/PARP5b inhibitor in vitro auto-PARsylation assay.