Learn More
Aviva Systems Biology Corp IWS1 Antibody - N-terminal region (ARP65333_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65333_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IWS1. It was validated on Western Blot. . Alternate names: DKFZp761G0123, FLJ10006, FLJ14655, FLJ32319, MGC126375, MGC126376ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-476