Learn More
Molecular Devices Molecular Devices Automated Cell Imaging System, Compact, Fluorescence, Digital Confocal
Catalog No. 5000014680
Encompass_Preferred
The ImageXpress Pico Automated Cell Imaging System is a compact, automated imaging and analysis platform designed to accelerate routine cell biology workflows. It combines high-resolution capture with onboard analysis to support brightfield, fluorescence, and confocal-style imaging for assay development and high-throughput experiments.
  • Compact form factor for benchtop use.
  • Automated image acquisition for high-throughput workflows.
  • Supports brightfield, fluorescence, and digital confocal imaging modes.
  • Integrated image analysis for rapid data extraction.
  • Optional environmental control for live-cell imaging.
  • Simplified workflow suited to assay development and screening.
Catalog No. 50-000-14680 Supplier Molecular Devices Supplier No. PICOADVG
Add to Cart
Add to Cart
Request a Quote
The ImageXpress Pico Automated Cell Imaging System is a compact, automated imaging and analysis platform designed to accelerate routine cell biology workflows. It combines high-resolution capture with onboard analysis to support brightfield, fluorescence, and confocal-style imaging for assay development and high-throughput experiments.
  • Compact form factor for benchtop use.
  • Automated image acquisition for high-throughput workflows.
  • Supports brightfield, fluorescence, and digital confocal imaging modes.
  • Integrated image analysis for rapid data extraction.
  • Optional environmental control for live-cell imaging.
  • Simplified workflow suited to assay development and screening.