Learn More
Molecular Devices Molecular Devices ImageXpress Pico, 4x to 63x, Fluorescence, Brightfield
Catalog No. 5000014672
Encompass_Preferred
The ImageXpress Pico Automated Cell Imaging System provides compact, automated benchtop imaging and integrated analysis for rapid cell-based assays. It combines high-resolution brightfield and fluorescence imaging with digital confocal deconvolution and supports plate- and slide-based workflows with optional environmental control for live-cell experiments.
  • Automated brightfield, fluorescence, and digital confocal imaging.
  • User-exchangeable long-working-distance objectives, 4x to 63x.
  • Icon-driven acquisition and analysis software with preconfigured protocols.
  • Supports 6- to 384-well plates, slides, and 35 mm dishes.
  • Environmental control options for humidity, CO2, and O2 during time-lapse experiments.
  • Autofocus, z-stack acquisition, and 6-position automated objective turret for high throughput and image quality.
Catalog No. 50-000-14672 Supplier Molecular Devices Supplier No. PICOBASICG
Add to Cart
Add to Cart
Request a Quote
The ImageXpress Pico Automated Cell Imaging System provides compact, automated benchtop imaging and integrated analysis for rapid cell-based assays. It combines high-resolution brightfield and fluorescence imaging with digital confocal deconvolution and supports plate- and slide-based workflows with optional environmental control for live-cell experiments.
  • Automated brightfield, fluorescence, and digital confocal imaging.
  • User-exchangeable long-working-distance objectives, 4x to 63x.
  • Icon-driven acquisition and analysis software with preconfigured protocols.
  • Supports 6- to 384-well plates, slides, and 35 mm dishes.
  • Environmental control options for humidity, CO2, and O2 during time-lapse experiments.
  • Autofocus, z-stack acquisition, and 6-position automated objective turret for high throughput and image quality.