Learn More
Medchemexpress LLC HY-139214 5mg , IXA4 CAS:1185329-96-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1392145MG
Encompass_Preferred
HY-139214 5mg IXA4 CAS: 1185329-96-7 IXA4 is a highly selective, non-toxic IRE1/XBP1s activator. IXA4 activates IRE1/XBP1s signaling without globally activating the unfolded protein response (UPR) or other stress-responsive signaling pathways (e.g., the heat shock response or oxidative stress response). IXA4 reduces secretion of APP through IRE1 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9357