Learn More
TARGETMOL CHEMICALS INC IXAZOMIB 10MG
SDP
Catalog No. 502577221
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ixazomib (MLN2238) a second generation boron-containing peptide proteasome inhibitor (PI) inhibits the chymotrypsin-like proteolytic ((beta)5) site of the 20S proteasome (IC50/Ki 3.4/0.93 nM in cell-free assays) also inhibits the caspase-like ((beta)1) and trypsin-like ((beta)2) proteolytic sites (IC50 31/3500 nM). purity: 98%
Catalog No. 50-257-7221 Supplier TARGETMOL CHEMICALS INC Supplier No. T212210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ixazomib (MLN2238) a second generation boron-containing peptide proteasome inhibitor (PI) inhibits the chymotrypsin-like proteolytic ((beta)5) site of the 20S proteasome (IC50/Ki 3.4/0.93 nM in cell-free assays) also inhibits the caspase-like ((beta)1) and trypsin-like ((beta)2) proteolytic sites (IC50 31/3500 nM). purity: 98%