Learn More
Selleck Chemical LLC Ixazomib (MLN2238) S2180-50mg
SDP
Catalog No. 503112320
Encompass_Preferred
Ixazomib (MLN2238) inhibits the chymotrypsin-like proteolytic ( 5) site of the 20S proteasome with IC50 and Ki of 3 4 nM and 0 93 nM in cell-free assays respectively also inhibits the caspase-like ( 1) and trypsin-like ( 2) proteolytic sites with IC50 of 31 and 3500 nM Ixazomib (MLN2238) induces autophagy Phase 3
Catalog No. 50-311-2320 Supplier Selleck Chemical LLC Supplier No. S218050MG
Ixazomib (MLN2238) inhibits the chymotrypsin-like proteolytic ( 5) site of the 20S proteasome with IC50 and Ki of 3 4 nM and 0 93 nM in cell-free assays respectively also inhibits the caspase-like ( 1) and trypsin-like ( 2) proteolytic sites with IC50 of 31 and 3500 nM Ixazomib (MLN2238) induces autophagy Phase 3