Learn More
Medchemexpress LLC HY-10453 25mg , Ixazomib CAS:1072833-77-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1045325MG
Encompass_Preferred
HY-10453 25mg Ixazomib CAS: 1072833-77-2 Ixazomib (MLN2238) is a selective, potent, and reversible proteasome inhibitor, which inhibits the chymotrypsin-like proteolytic (β5) site of the 20S proteasome with an IC50 of 3.4 nM (Ki of 0.93 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0468