Learn More
Selleck Chemical LLC Ixazomib Citrate (MLN9708) S4432-25mg
SDP
Catalog No. 503109493
Encompass_Preferred
Ixazomib Citrate (MLN9708) is a prodrug of Ixazomib (MLN2238) which is a selective orally bioavailable inhibitor of 20S proteasome that inhibits the chymotrypsin-like proteolytic ( 5) site with IC50 of 3 4 nM and Ki of 0 93 nM respectively Ixazomib (MLN2238) also inhibits caspase-like ( 1) and trypsin-like ( 2) proteolytic sites with IC50 of 31 nM and 3500 nM respectively
Catalog No. 50-310-9493 Supplier Selleck Chemical LLC Supplier No. S443225MG
Ixazomib Citrate (MLN9708) is a prodrug of Ixazomib (MLN2238) which is a selective orally bioavailable inhibitor of 20S proteasome that inhibits the chymotrypsin-like proteolytic ( 5) site with IC50 of 3 4 nM and Ki of 0 93 nM respectively Ixazomib (MLN2238) also inhibits caspase-like ( 1) and trypsin-like ( 2) proteolytic sites with IC50 of 31 nM and 3500 nM respectively