Learn More
AdipoGen Ixazomib MLN2238
SDP
Catalog No. 502562241
Encompass_Preferred
Chemical. CAS 1072833-77-2. Formula C14H19BCl2N2O4. MW 361. Synthetic. Potent selective and reversible proteasome inhibitor all proteolytic subunits. Targets the chymotrypsin-like beta5 subunit of the constitutive 20S proteasome IC50=3.4nM. Cross-reacts and inhibits the trypsin-like beta2 subunit IC50=3.5µM and the caspase-like/peptidyl-glutamyl peptide-hydrolyzing PGPH beta1 subunit IC50=0.03µM. Anticancer compound effective in cell-based assays, in xenografts and against multiple myeloma in vivo. In vitro, induces cell cycle arrest and apoptosis in human cancer cell lines including multiple myeloma. Biologically active form of the prodrug MLN9708 AG-CR1-3671. Exhibits improved pharmacodynamics and antitumor activity compared to bortezomib in various B-cell lymphoma models, due to a greater tumor to blood ratio of proteasome inhibition that ultimately translates into improved tumor pharmacodynamic response and antitumor activity in several tumor xenograft models.
Catalog No. 50-256-2241 Supplier AdipoGen Supplier No. AGCR13670M005
Chemical. CAS 1072833-77-2. Formula C14H19BCl2N2O4. MW 361. Synthetic. Potent selective and reversible proteasome inhibitor all proteolytic subunits. Targets the chymotrypsin-like beta5 subunit of the constitutive 20S proteasome IC50=3.4nM. Cross-reacts and inhibits the trypsin-like beta2 subunit IC50=3.5µM and the caspase-like/peptidyl-glutamyl peptide-hydrolyzing PGPH beta1 subunit IC50=0.03µM. Anticancer compound effective in cell-based assays, in xenografts and against multiple myeloma in vivo. In vitro, induces cell cycle arrest and apoptosis in human cancer cell lines including multiple myeloma. Biologically active form of the prodrug MLN9708 AG-CR1-3671. Exhibits improved pharmacodynamics and antitumor activity compared to bortezomib in various B-cell lymphoma models, due to a greater tumor to blood ratio of proteasome inhibition that ultimately translates into improved tumor pharmacodynamic response and antitumor activity in several tumor xenograft models.