Learn More
Medchemexpress LLC HY-P9924 5mg , Ixekizumab CAS:1143503-69-8 Purity:>98%
SDP
Catalog No. 502261221
Encompass_Preferred
HY-P9924 5mg Ixekizumab CAS: 1143503-69-8 Ixekizumab (LY2439821) is a humanized IgG4 monoclonal antibody that selectively binds and neutralizes interleukin IL-17A (KD<3 pM). Ixekizumab directly blocks IL-17A binding to IL-17RA (IL-17A receptor) but does not bind to other IL-17 family members. Ixekizumab is used for the research of moderate-to-severe plaque psoriasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1221 Supplier Medchemexpress LLC Supplier No. HYP99245MG
HY-P9924 5mg Ixekizumab CAS: 1143503-69-8 Ixekizumab (LY2439821) is a humanized IgG4 monoclonal antibody that selectively binds and neutralizes interleukin IL-17A (KD<3 pM). Ixekizumab directly blocks IL-17A binding to IL-17RA (IL-17A receptor) but does not bind to other IL-17 family members. Ixekizumab is used for the research of moderate-to-severe plaque psoriasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.