Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IYPTNGYTR (acetate) | 99.4% | 1144.23 | 10 MG
SDP

Catalog No. 5000399310
Encompass_Preferred

This product is IYPTNGYTR acetate, a deamidation-sensitive signature peptide used to monitor in vivo Trastuzumab metabolism. It is classified as a laboratory chemical for research use only and is not considered a hazardous substance or mixture. It appears as a solid. Recommended storage for powder is -80°C for 2 years or -20°C for 1 year. If in solvent, store at -80°C for 6 months or -20°C for 1 month in sealed conditions away from moisture.

  • Deamidation-sensitive signature peptide
  • Used to monitor in vivo Trastuzumab metabolism
  • For research use only
  • Not classified as a hazardous substance or mixture
  • Appears as a solid
  • Powder storage: -80°C for 2 years; -20°C for 1 year
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-003-99310 Supplier Medchemexpress LLC Supplier No. HYP3147A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is IYPTNGYTR acetate, a deamidation-sensitive signature peptide used to monitor in vivo Trastuzumab metabolism. It is classified as a laboratory chemical for research use only and is not considered a hazardous substance or mixture. It appears as a solid. Recommended storage for powder is -80°C for 2 years or -20°C for 1 year. If in solvent, store at -80°C for 6 months or -20°C for 1 month in sealed conditions away from moisture.

  • Deamidation-sensitive signature peptide
  • Used to monitor in vivo Trastuzumab metabolism
  • For research use only
  • Not classified as a hazardous substance or mixture
  • Appears as a solid
  • Powder storage: -80°C for 2 years; -20°C for 1 year
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.