Learn More
Medchemexpress LLC HY-111411 5mg Medchemexpress, IZCZ-3 CAS:2223019-53-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1114115MG
Encompass_Preferred
Medchemexpress, HY-111411 5mg IZCZ-3 CAS:2223019-53-0 IZCZ-3 is a potent c-MYC transcription inhibitor with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9694