Learn More
AdipoGen izTRAIL, Soluble (human) (rec.) (5 x 10 µg)
SDP
Supplier:  AdipoGen AG40B00695010
Encompass_Preferred
Protein. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to an isoleucine zipper motif. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains 20mM TRIS-Cl, 0.5M arginine-HCl, 100mM NaCl, 0.02% Tween 20. Binds to human TRAIL receptors 1-4 (TRAIL-R1 to TRAIL-R4); izTRAIL does not interact with the apoptosis-inducing mouse TRAIL receptor (TRAIL-R). Purity: >95% (SDS-PAGE). izTRAIL is a newly available, highly active recombinant form of soluble human TRAIL. Due to a trimerizing N-terminal isoleucine zipper (iz) motif the intrinsic trimerization of TRAIL, required for apoptosis-inducing activity of TRAIL, is enhanced when compared to non-tagged soluble human TRAIL (shTRAIL). Therefore, izTRAIL is a potent inducer of apoptosis in many human cancer cells, but not normal human hepatocytes. In addition, the half-life of izTRAIL is about eight-fold higher than the half-life of shTRAIL.
Catalog No. 50-205-3125