Learn More
Aviva Systems Biology Corp IZUMO1 Antibody - C-terminal region (ARP51341_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51341_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IZUMO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human IZUMO1. Alternate names: IZUMO, MGC34799ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60553