Learn More
Medchemexpress LLC J-113863 | 353791-85-2 | MFCD09971097 | ≥99.0% | 655.44 | C30H37Cl2IN2O2 | 5 MG
SDP
Catalog No. 5000244893
Encompass_Preferred
J-113863 is a small-molecule chemokine receptor antagonist selective for CCR1, with demonstrated activity against human CCR3. Supplied as a solid research reagent (CAS 353791-85-2), it is intended for in vitro and preclinical receptor pharmacology studies.
  • Potent CCR1 antagonist activity (human IC50 0.9 nM).
  • Also active at human CCR3 (IC50 0.58 nM).
  • Provided as small solid quantities suitable for biochemical and cell-based assays.
  • Supplied with COA and SDS for lot-specific quality and handling information.
  • Storage recommendations available for solid and solution formats to preserve stability.
Catalog No. 50-002-44893 Supplier Medchemexpress LLC Supplier No. HY1033605MG
J-113863 is a small-molecule chemokine receptor antagonist selective for CCR1, with demonstrated activity against human CCR3. Supplied as a solid research reagent (CAS 353791-85-2), it is intended for in vitro and preclinical receptor pharmacology studies.
  • Potent CCR1 antagonist activity (human IC50 0.9 nM).
  • Also active at human CCR3 (IC50 0.58 nM).
  • Provided as small solid quantities suitable for biochemical and cell-based assays.
  • Supplied with COA and SDS for lot-specific quality and handling information.
  • Storage recommendations available for solid and solution formats to preserve stability.