Learn More
Medchemexpress LLC J-113863 | 353791-85-2 | MFCD09971097 | ≥99.0% | 655.44 g/mol | C30H37Cl2IN2O2 | 100 MG
SDP
Catalog No. 5000245210
Encompass_Preferred
J-113863 is a potent, selective small-molecule antagonist of C-C chemokine receptor 1 (CCR1) used in research on chemokine signaling and inflammation (CAS 353791-85-2). It exhibits sub-nanomolar to low-nanomolar activity versus CCR1 and potent activity versus human CCR3.
  • Potent CCR1 antagonist; human CCR1 IC50 = 0.9 nM, mouse CCR1 IC50 = 5.8 nM.
  • Potent human CCR3 antagonist (IC50 = 0.58 nM).
  • Available as solid powder and as a DMSO solution; multiple pack sizes including 100 mg.
  • High purity (≥99.0%) suitable for in vitro pharmacology.
  • Used in chemokine receptor and inflammation research applications.
Catalog No. 50-002-45210 Supplier Medchemexpress LLC Supplier No. HY103360100MG
J-113863 is a potent, selective small-molecule antagonist of C-C chemokine receptor 1 (CCR1) used in research on chemokine signaling and inflammation (CAS 353791-85-2). It exhibits sub-nanomolar to low-nanomolar activity versus CCR1 and potent activity versus human CCR3.
  • Potent CCR1 antagonist; human CCR1 IC50 = 0.9 nM, mouse CCR1 IC50 = 5.8 nM.
  • Potent human CCR3 antagonist (IC50 = 0.58 nM).
  • Available as solid powder and as a DMSO solution; multiple pack sizes including 100 mg.
  • High purity (≥99.0%) suitable for in vitro pharmacology.
  • Used in chemokine receptor and inflammation research applications.