Learn More
MSC Diversey All-Purpose Cleaner, Liquid Concentrate, Hanging Bag, 2.5 gal
Catalog No. 502817378
Encompass
This all-purpose cleaner is a liquid concentrate designed for commercial applications. It comes in a 2.5 gal hanging bag and is suitable for cleaning hard, non-porous surfaces in settings like commercial sinks, bars, institutions, dairies, and restaurants. It is compliant with NSF, Halal, and Kosher standards.
  • No need to rinse; allow to drain and air dry
  • Operates in a 200 to 400 ppm range
  • Liquid concentrate form
  • Typical quaternary scent
  • Recommended dilution: 1:512
  • VOC content: 1%
Catalog No. 50-281-7378 Supplier MSC Supplier No. 03008869
This all-purpose cleaner is a liquid concentrate designed for commercial applications. It comes in a 2.5 gal hanging bag and is suitable for cleaning hard, non-porous surfaces in settings like commercial sinks, bars, institutions, dairies, and restaurants. It is compliant with NSF, Halal, and Kosher standards.
  • No need to rinse; allow to drain and air dry
  • Operates in a 200 to 400 ppm range
  • Liquid concentrate form
  • Typical quaternary scent
  • Recommended dilution: 1:512
  • VOC content: 1%