Learn More
MSC Diversey J-512 Sanitizer, No-Rinse, Food Contact, 2.5 L, 2/Carton
Catalog No. 502817140
Encompass
The Diversey J-512 Sanitizer is a no-rinse sanitizer designed for effectively sanitizing food contact surfaces where hygiene is critical. It is suitable for use in dairies, restaurants, bars, and institutional settings.
  • Sanitizes food contact and other hard, nonporous environmental surfaces
  • No need to rinse; simply allow to drain and air dry
  • Effective within a PPM range of 200–400 ppm
  • Can be used in commercial sinks for immersion of utensils
  • Features a pink color with a quaternary scent
  • Catalog dilution ratio range of 1:512
Catalog No. 50-281-7140 Supplier MSC Supplier No. 03055225
The Diversey J-512 Sanitizer is a no-rinse sanitizer designed for effectively sanitizing food contact surfaces where hygiene is critical. It is suitable for use in dairies, restaurants, bars, and institutional settings.
  • Sanitizes food contact and other hard, nonporous environmental surfaces
  • No need to rinse; simply allow to drain and air dry
  • Effective within a PPM range of 200–400 ppm
  • Can be used in commercial sinks for immersion of utensils
  • Features a pink color with a quaternary scent
  • Catalog dilution ratio range of 1:512