Learn More
MSC Diversey Dispensing System, Four Dispenser, Safe Gap, Wall Mounted
Catalog No. 502839577
Encompass
This wall-mounted chemical dispensing system accurately dispenses four products into bottles, buckets, or floor care machines. It features SafeGap and Air Gap eductors for ASSE 1055B backflow prevention, using super-concentrated chemicals to reduce costs and waste.
  • Rugged steel doors, lockable cabinet doors, and product identification windows.
  • Drip tray with drain hose; remote high-flow bucket/auto scrubber fill gun.
  • One-handed 1.5 GPM low-flow and low-foam bottle fill.
  • 8 ft. 3.1 GPM high-flow bucket fill hose with easy point-of-use activation.
  • Smartvalve technology automatically chooses flow rate.
  • SafeGap and Air Gap eductors meet ASSE 1055B standard for backflow prevention.
  • Uses super-concentrated chemicals, reducing in-use cost and waste.
  • Leak-proof Spill-Tite cartridges eliminate chemical exposure, tampering, and manual dilution.
Catalog No. 50-283-9577 Supplier MSC Supplier No. 88209077
This wall-mounted chemical dispensing system accurately dispenses four products into bottles, buckets, or floor care machines. It features SafeGap and Air Gap eductors for ASSE 1055B backflow prevention, using super-concentrated chemicals to reduce costs and waste.
  • Rugged steel doors, lockable cabinet doors, and product identification windows.
  • Drip tray with drain hose; remote high-flow bucket/auto scrubber fill gun.
  • One-handed 1.5 GPM low-flow and low-foam bottle fill.
  • 8 ft. 3.1 GPM high-flow bucket fill hose with easy point-of-use activation.
  • Smartvalve technology automatically chooses flow rate.
  • SafeGap and Air Gap eductors meet ASSE 1055B standard for backflow prevention.
  • Uses super-concentrated chemicals, reducing in-use cost and waste.
  • Leak-proof Spill-Tite cartridges eliminate chemical exposure, tampering, and manual dilution.