Learn More
Chemglass Life Sciences J-Kem 150-J Temperature Controller Only, "J" Type
SDP
Catalog No. CG319811
Encompass_Preferred
J-Kem 150-J Temperature Controller Only, "J". Type THERMOCOUPLE TYPE (TEMPERATURE RANGE):T (-200 to 250¿C), J (0 to 800¿C), K (-50 to 1200¿C)
Catalog No. CG319811 Supplier Chemglass Life Sciences Supplier No. CG319811
J-Kem 150-J Temperature Controller Only, "J". Type THERMOCOUPLE TYPE (TEMPERATURE RANGE):T (-200 to 250¿C), J (0 to 800¿C), K (-50 to 1200¿C)