Learn More
Chemglass Life Sciences J-Kem 150-T-S Temperature Controller, Complete System, "T" Type
SDP
Catalog No. CG319801
Encompass_Preferred
J-Kem 150-T-S Temperature Controller, Complete System,"T" Type. THERMOCOUPLE TYPE (TEMPERATURE RANGE):T (-200 to 250¿C), J (0 to 800¿C), K (-50 to 1200¿C)
Catalog No. CG319801 Supplier Chemglass Life Sciences Supplier No. CG319801
J-Kem 150-T-S Temperature Controller, Complete System,"T" Type. THERMOCOUPLE TYPE (TEMPERATURE RANGE):T (-200 to 250¿C), J (0 to 800¿C), K (-50 to 1200¿C)