Learn More
Chemglass Life Sciences J-Kem 150-T Temperature Controller Only, "T" Type
SDP
Catalog No. CG319810
Encompass_Preferred
J-Kem 150-T Temperature Controller Only, "T" Type THERMOCOUPLE TYPE (TEMPERATURE RANGE):T (-200 to 250¿C), J (0 to 800¿C), K (-50 to 1200¿C)
Catalog No. CG319810 Supplier Chemglass Life Sciences Supplier No. CG319810
J-Kem 150-T Temperature Controller Only, "T" Type THERMOCOUPLE TYPE (TEMPERATURE RANGE):T (-200 to 250¿C), J (0 to 800¿C), K (-50 to 1200¿C)