Learn More
Chemglass Life Sciences J-Kem Temperature Controller, Model 210, Type "K" (-50°C to 1200°C), Complete
SDP
Supplier:  Chemglass Life Sciences CG320003
Encompass_Preferred
J-Kem Temperature Controller, Model 210, Type "K" (-50C to 1200C), Complete. The Model 210 has 1200 Watts of power which is sufficient for heating mantles up to 5L in size along with most laboratory ovens and hot plates. Controller has the ability to display temperature without heating. The Model 210 controls temperature to within 0.1C of liquids from 1ul to 5L.PLEASE NOTE: THE TYPE T, J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 3.25" H x 5.25" W x 7.25" DPower: 10 Amps @ 120VAC, 1200 Watts
Catalog No. CG320003