Learn More
Chemglass Life Sciences J-Kem Temperature Controller, Model 210/T, Type "K" (-50°C to 1200°C), Complete
SDP
Catalog No. CG320203
Encompass_Preferred
J-Kem Temperature Controller, Model 210/T, Type "K" (-50C to 1200C), Complete. Similar to CG-3200, but with the addition of a 100 hour digital timer to turn heating ON or OFF when time has expired.PLEASE NOTE: THE TYPE T,J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 3.4" H x 6.75" W x 6" DPower: 10 Amps @ 120VAC, 840 Watts
Catalog No. CG320203 Supplier Chemglass Life Sciences Supplier No. CG320203
J-Kem Temperature Controller, Model 210/T, Type "K" (-50C to 1200C), Complete. Similar to CG-3200, but with the addition of a 100 hour digital timer to turn heating ON or OFF when time has expired.PLEASE NOTE: THE TYPE T,J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 3.4" H x 6.75" W x 6" DPower: 10 Amps @ 120VAC, 840 Watts