Learn More
Chemglass Life Sciences Thin Wire Thermocouple, PTFE Coated, 36" Long, Type "K" (YELLOW)
SDP
Catalog No. CG321204
Encompass_Preferred
Thin Wire Thermocouple, PTFE Coated, 36" Long, Type "K" (YELLOW). Thin wire thermocouple is 30-gauge x 36" long, completely encased in PTFE for submersion in reaction. Excellent for use with small volumes, it can be inserted through rubber septa.
Catalog No. CG321204 Supplier Chemglass Life Sciences Supplier No. CG321204
Thin Wire Thermocouple, PTFE Coated, 36" Long, Type "K" (YELLOW). Thin wire thermocouple is 30-gauge x 36" long, completely encased in PTFE for submersion in reaction. Excellent for use with small volumes, it can be inserted through rubber septa.