Learn More
Sigma Aldrich Fine Chemicals Biosciences J KEMR digital vacuum regu1EA
Catalog No. 502908916
Encompass
Solid-state with digital pressure entry and displayWetted parts are SS with built-in SS vacuum sensor and valveConnects to standard vacuum pumps or any vacuum source and any piece of laboratory equipment to regulate its pressure to /-1 torr (mm Hg)Display accuracy is 0.3 % of rangePressure range 0.1 to 760.0 Torr (mm Hg)Both USB 2.0 and RS232 communication ports for PC control and data loggingFree windows software for PC control and data logging to Excel or text file. Implements multi-step temperature ramping and real-time strip chart displayUnit measures pressure in the attached piece of equipment and compares it to the desired pressure entered into the regulators digital display then opens or closes a valve between equipment and vacuum source to maintain desired pressure.A SS needle valve is suggested for use with the vacuum regulator and is useful to limit the rate of evacuation when the regulator is attached to small reactors or other pieces o
Catalog No. 50-290-8916 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2481691EA
Solid-state with digital pressure entry and displayWetted parts are SS with built-in SS vacuum sensor and valveConnects to standard vacuum pumps or any vacuum source and any piece of laboratory equipment to regulate its pressure to /-1 torr (mm Hg)Display accuracy is 0.3 % of rangePressure range 0.1 to 760.0 Torr (mm Hg)Both USB 2.0 and RS232 communication ports for PC control and data loggingFree windows software for PC control and data logging to Excel or text file. Implements multi-step temperature ramping and real-time strip chart displayUnit measures pressure in the attached piece of equipment and compares it to the desired pressure entered into the regulators digital display then opens or closes a valve between equipment and vacuum source to maintain desired pressure.A SS needle valve is suggested for use with the vacuum regulator and is useful to limit the rate of evacuation when the regulator is attached to small reactors or other pieces o