Learn More
Sigma Aldrich Fine Chemicals Biosciences J KEMR dual temperature co1EA
Catalog No. 502908934
Encompass
Research grade temperature controller features auto-tune PID process controllers with solid-state output circuitry and J-KEMs Power Control Computer that virtually eliminates temperature overshoot of the set temperature. Regulates any piece of equipment or volume to 0.1C.Regulates heating mantles hot plates ovens heating blocks etcSafe 100% solid-state electronics double fused unbreakable thermocouple probe automatic over-temperature circuitProgrammable ramp-to-set point temperature control on the 200-series modelFree windows software for PC control and data logging to Excel or text file. Implements multi-step temperature ramping and real-time strip chart displayBoth USB 2.0 and RS232 communication ports for PC control and data loggingGemini Powers two independent reactions with 1 200 W per channelDual temperature display100 h digital timer on channel 1Programmable for heating and coolingOver-temperature circuit stops heating an
Catalog No. 50-290-8934 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2800701EA
Research grade temperature controller features auto-tune PID process controllers with solid-state output circuitry and J-KEMs Power Control Computer that virtually eliminates temperature overshoot of the set temperature. Regulates any piece of equipment or volume to 0.1C.Regulates heating mantles hot plates ovens heating blocks etcSafe 100% solid-state electronics double fused unbreakable thermocouple probe automatic over-temperature circuitProgrammable ramp-to-set point temperature control on the 200-series modelFree windows software for PC control and data logging to Excel or text file. Implements multi-step temperature ramping and real-time strip chart displayBoth USB 2.0 and RS232 communication ports for PC control and data loggingGemini Powers two independent reactions with 1 200 W per channelDual temperature display100 h digital timer on channel 1Programmable for heating and coolingOver-temperature circuit stops heating an