Learn More
Sigma Aldrich Fine Chemicals Biosciences J KEMR safety temperature1EA
Catalog No. 502909076
Encompass
Designed for processes requiring uncompromised safety. Containsboth a digital temperature controller and an independent back up controller in the same cabinet to provide 100% redundant control of safety critical reactions. The set temperature is entered into the primary digital meter then a high temperature limit is entered into the backup digital meter. Both meters monitor the reaction and in the event of an electrical failure or over temperature condition the remaining meter takes control and permanently turns off output power.Regulates heating mantles hot plates ovens heating blocks etc.Safe 100% solid-state electronics double fused unbreakable thermocouple probe automatic over-temperature circuitProgrammable ramp-to-set point temperature control on the 200-Series modelSerial communications for PC control and data loggingTwo power outlets for large heaters and mantles up to 22 LOver-temperature circuit stops heating and sounds alarmBoth US
Catalog No. 50-290-9076 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2482741EA
Designed for processes requiring uncompromised safety. Containsboth a digital temperature controller and an independent back up controller in the same cabinet to provide 100% redundant control of safety critical reactions. The set temperature is entered into the primary digital meter then a high temperature limit is entered into the backup digital meter. Both meters monitor the reaction and in the event of an electrical failure or over temperature condition the remaining meter takes control and permanently turns off output power.Regulates heating mantles hot plates ovens heating blocks etc.Safe 100% solid-state electronics double fused unbreakable thermocouple probe automatic over-temperature circuitProgrammable ramp-to-set point temperature control on the 200-Series modelSerial communications for PC control and data loggingTwo power outlets for large heaters and mantles up to 22 LOver-temperature circuit stops heating and sounds alarmBoth US