Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc J SPHERE H80 4UM 80A 50X2MM
SDP

Catalog No. 50157006
Encompass_Preferred

JH08S04-0502WT: J'Sphere H80, 4um,; 80A, 50X2mm Column

Catalog No. 50-157-006 Supplier YMC America, Inc Supplier No. JH08S040502WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JH08S04-0502WT: J'Sphere H80, 4um,; 80A, 50X2mm Column

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.