Learn More
Chemglass Life Sciences J-Kem Temperature Controller, Model 250, Type "J" (0°C to 800°C), Complete
SDP
Catalog No. CG320402
Encompass_Preferred
J-Kem Temperature Controller, Model 250, Type "J" (0C to 800C), Complete. The Model 250 has 3 power outlets for maximum versatility in research. Two of the outlets are used for heating up to 1800 watts (mantles up to 22L).The third outlet (360 watts), used normally for cooling, is programmable to provide power below, above, or at the entered temperature. This third outlet can also be used to turn on an alarm for an over or under temperature condition. Controller has the ability to display temperature without heating. The Model 250 controls temperature to within 0.1C of liquids from 1ul to 22L. Available w/ a choice of Type "T", J or K thermocouple.PLEASE NOTE: CONTROLLERS ARE SUPPLIED COMPLETE WITH 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. Dimensions: 3.25" H x 7.5" W x 9" DPower: Heating Outlets #1 & #2: 15 Amps @ 120VAC, 1800 Watts Cooling/Alarm Outlet #3: 3 Amps @ 12VAC, 360 Watts
Catalog No. CG320402 Supplier Chemglass Life Sciences Supplier No. CG320402
J-Kem Temperature Controller, Model 250, Type "J" (0C to 800C), Complete. The Model 250 has 3 power outlets for maximum versatility in research. Two of the outlets are used for heating up to 1800 watts (mantles up to 22L).The third outlet (360 watts), used normally for cooling, is programmable to provide power below, above, or at the entered temperature. This third outlet can also be used to turn on an alarm for an over or under temperature condition. Controller has the ability to display temperature without heating. The Model 250 controls temperature to within 0.1C of liquids from 1ul to 22L. Available w/ a choice of Type "T", J or K thermocouple.PLEASE NOTE: CONTROLLERS ARE SUPPLIED COMPLETE WITH 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. Dimensions: 3.25" H x 7.5" W x 9" DPower: Heating Outlets #1 & #2: 15 Amps @ 120VAC, 1800 Watts Cooling/Alarm Outlet #3: 3 Amps @ 12VAC, 360 Watts