Learn More
Hach Company Tube for General Purpose Distillation Apparatus, round bottom, 500 mL
Catalog No. 501995670 Shop All Hach Company Products
Encompass
Designed for use with the General Purpose Distillation Apparatus. Round bottom, 500 mL capacity. Long neck with branch and thermometer port. Neck extends 90 mm above flask body and branch extends 150 mm above flask body. Both neck and branch have 20-400 screw-thread finish. Thermometer port has 13-425 screw-thread finish.
Catalog No. 50-199-5670 Supplier Hach Company Supplier No. 2264900
Designed for use with the General Purpose Distillation Apparatus. Round bottom, 500 mL capacity. Long neck with branch and thermometer port. Neck extends 90 mm above flask body and branch extends 150 mm above flask body. Both neck and branch have 20-400 screw-thread finish. Thermometer port has 13-425 screw-thread finish.