Learn More
Agilent Technologies J&W DB-624 ULTRA INERT GC COLU
Catalog No. NC2240852
Encompass
J&W DB-624 Ultra Inert GC Column, 60 m, 0.32 mm, 1.80 um, 7 inch cage
This item is not returnable. View return policy
Catalog No. NC2240852 Supplier Agilent Technologies Supplier No. 1231364UI
J&W DB-624 Ultra Inert GC Column, 60 m, 0.32 mm, 1.80 um, 7 inch cage