Learn More
MSC J.W. Winco Fluid Sight Glass, Aluminum, ESG Safety Glass, M33x1.5
Catalog No. 5000512050
Encompass
Fluid sight glasses are designed for high stability and scratch resistance. Their unique seal placement on the outer diameter of the glass prevents leakage, regardless of tightening. The embedded seal in a radial recess ensures it cannot be lost or squeezed out during installation. These sight glasses are suitable for use on pressurized tanks, offering reliable performance.
  • Scratch-proof sight glass of high stability
  • Leakage eliminated by seal placement on outer diameter
  • Seal is embedded in a radial recess to prevent loss
  • Suitable for use on pressurized tanks
  • Resistant to temperatures up to 356 °F
  • Constructed with an aluminum body
  • Features ESG safety glass
  • Equipped with a reflector made from Polyethersulfone plastic
  • Includes a fluorine rubber (FKM) seal
  • RoHS compliant
Catalog No. 50-005-12050 Supplier MSC Supplier No. 64829369
Fluid sight glasses are designed for high stability and scratch resistance. Their unique seal placement on the outer diameter of the glass prevents leakage, regardless of tightening. The embedded seal in a radial recess ensures it cannot be lost or squeezed out during installation. These sight glasses are suitable for use on pressurized tanks, offering reliable performance.
  • Scratch-proof sight glass of high stability
  • Leakage eliminated by seal placement on outer diameter
  • Seal is embedded in a radial recess to prevent loss
  • Suitable for use on pressurized tanks
  • Resistant to temperatures up to 356 °F
  • Constructed with an aluminum body
  • Features ESG safety glass
  • Equipped with a reflector made from Polyethersulfone plastic
  • Includes a fluorine rubber (FKM) seal
  • RoHS compliant