Learn More
MSC J.W. Winco Fluid Sight Glass, Aluminum, G 1 Thread, 36 mm Distance Across Flats
Catalog No. 5000531033
Encompass
J.W. Winco Fluid Sight Glasses GN 743 provide stable and reliable fluid level monitoring. They feature a scratch-proof sight glass and a unique, leak-preventing seal placed on the outer diameter. Designed with a recessed hex for DIN 3852 pipe fittings, their embedded seal ensures secure installation without loss or squeezing. These sight glasses are also suitable for pressurized tank applications.
  • Scratch-proof sight glass for high stability.
  • Unique seal placement prevents leakage.
  • Recessed hex fits DIN 3852 pipe fittings.
  • Embedded seal ensures secure installation.
  • Suitable for pressurized tanks.
  • Aluminum body and float glass construction.
  • Operating temperature up to 212 °F (100 °C).
  • RoHS compliant.
Catalog No. 50-005-31033 Supplier MSC Supplier No. 64829187
J.W. Winco Fluid Sight Glasses GN 743 provide stable and reliable fluid level monitoring. They feature a scratch-proof sight glass and a unique, leak-preventing seal placed on the outer diameter. Designed with a recessed hex for DIN 3852 pipe fittings, their embedded seal ensures secure installation without loss or squeezing. These sight glasses are also suitable for pressurized tank applications.
  • Scratch-proof sight glass for high stability.
  • Unique seal placement prevents leakage.
  • Recessed hex fits DIN 3852 pipe fittings.
  • Embedded seal ensures secure installation.
  • Suitable for pressurized tanks.
  • Aluminum body and float glass construction.
  • Operating temperature up to 212 °F (100 °C).
  • RoHS compliant.