Learn More
MSC J.W. Winco Oil Level Sight Glass, Aluminum, M26x1.5 Thread, 30 mm Distance Across Flats
Catalog No. 5000531034
Encompass
This oil level sight glass is designed for accurately monitoring fluid levels. It features robust aluminum construction, an M26x1.5 thread for secure installation, and a durable glass window for clear visibility.
  • Aluminum body ensures durability
  • M26x1.5 thread for easy and secure fitting
  • 30 mm distance across flats for precise installation
  • 32 mm flange diameter
  • 8 mm head height
  • 9 mm length under head
  • Glass window provides clear visibility
  • RoHS compliant
Catalog No. 50-005-31034 Supplier MSC Supplier No. 64829278
This oil level sight glass is designed for accurately monitoring fluid levels. It features robust aluminum construction, an M26x1.5 thread for secure installation, and a durable glass window for clear visibility.
  • Aluminum body ensures durability
  • M26x1.5 thread for easy and secure fitting
  • 30 mm distance across flats for precise installation
  • 32 mm flange diameter
  • 8 mm head height
  • 9 mm length under head
  • Glass window provides clear visibility
  • RoHS compliant