Learn More
MSC J.W. Winco Oil Level Sight Glass, Aluminum, M33x1.5 Thread
Catalog No. 5000535405
Encompass
This oil level sight glass is designed with an M33x1.5 thread and features a 36 mm distance across flats. Constructed from aluminum with a glass window, it has a 40 mm flange diameter, an 8.5 mm head height, and an 11 mm length under the head, making it suitable for various industrial applications.
  • M33x1.5 thread for secure installation.
  • 36 mm distance across flats.
  • Durable aluminum construction with a clear glass window.
  • 40 mm flange diameter.
  • 8.5 mm head height.
  • 11 mm length under head.
  • RoHS compliant.
Catalog No. 50-005-35405 Supplier MSC Supplier No. 64829294
This oil level sight glass is designed with an M33x1.5 thread and features a 36 mm distance across flats. Constructed from aluminum with a glass window, it has a 40 mm flange diameter, an 8.5 mm head height, and an 11 mm length under the head, making it suitable for various industrial applications.
  • M33x1.5 thread for secure installation.
  • 36 mm distance across flats.
  • Durable aluminum construction with a clear glass window.
  • 40 mm flange diameter.
  • 8.5 mm head height.
  • 11 mm length under head.
  • RoHS compliant.