Learn More
MSC J.W. Winco Oil Level Sight Glass, M20x1.5 Thread, Aluminum
Catalog No. 5000514634
Encompass
This oil level sight glass features an M20x1.5 thread and is constructed with an aluminum body and a glass window, making it suitable for industrial fluid level monitoring applications.
  • M20x1.5 thread
  • 23 mm distance across flats
  • Aluminum body material
  • Glass window material
  • 26 mm flange diameter
  • 7.5 mm head height
  • 8.5 mm length under head
  • RoHS compliant
  • Country of origin: Germany
Catalog No. 50-005-14634 Supplier MSC Supplier No. 64829260
This oil level sight glass features an M20x1.5 thread and is constructed with an aluminum body and a glass window, making it suitable for industrial fluid level monitoring applications.
  • M20x1.5 thread
  • 23 mm distance across flats
  • Aluminum body material
  • Glass window material
  • 26 mm flange diameter
  • 7.5 mm head height
  • 8.5 mm length under head
  • RoHS compliant
  • Country of origin: Germany