Learn More
MSC J.W. Winco Oil Level Sight Glass, Plastic, G 3/4 Thread, 28 mm Distance Across Flats
Catalog No. 5000513880
Encompass
This oil level sight glass features a durable plastic window material and is designed with a G 3/4 thread. It has a 28 mm distance across flats, a 35 mm flange diameter, and an 11 mm length under the head. This product is suitable for various applications requiring precise fluid level monitoring.
  • RoHS compliant for environmental safety
  • Durable plastic construction for reliable performance
  • Designed for easy installation with a G 3/4 thread
  • Provides clear visibility for accurate fluid level monitoring
Catalog No. 50-005-13880 Supplier MSC Supplier No. 64829120
This oil level sight glass features a durable plastic window material and is designed with a G 3/4 thread. It has a 28 mm distance across flats, a 35 mm flange diameter, and an 11 mm length under the head. This product is suitable for various applications requiring precise fluid level monitoring.
  • RoHS compliant for environmental safety
  • Durable plastic construction for reliable performance
  • Designed for easy installation with a G 3/4 thread
  • Provides clear visibility for accurate fluid level monitoring