Learn More
Sigma Aldrich Fine Chemicals Biosciences J.Young high-vacuum PTfE v1EA
Catalog No. 502900353
Encompass
The J. Young high-vacuum PTfE valve helps in controlling the atmosphere when using air-sensitive materials.featuresPTfE piston with unique PTfE-sheathed elastomer O-ring sealsGrease-free glass-to-PTfE sealsHigh-vacuum to 10-6 torr
Catalog No. 50-290-0353 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4055901EA
The J. Young high-vacuum PTfE valve helps in controlling the atmosphere when using air-sensitive materials.featuresPTfE piston with unique PTfE-sheathed elastomer O-ring sealsGrease-free glass-to-PTfE sealsHigh-vacuum to 10-6 torr