Learn More
Grainger ARC Abrasives Predator Quick-Change Sanding Disc, 4 in., Ceramic, 36 Grit, 25/Pk.
Catalog No. 502772342
Encompass_Preferred
These quick-change sanding discs are designed for effective material removal and surface preparation. Featuring a high-performance ceramic abrasive on a durable Y weight cloth backing, these discs provide reliable performance for heavy-duty applications. The TR roll-on/off attachment system allows for fast and secure disc changes, minimizing downtime during operation.
  • Ceramic abrasive material provides durability and cutting efficiency
  • 36 grit extra coarse abrasive grade for aggressive material removal
  • 4 in. diameter for versatile surface work
  • Durable Y weight cloth backing for demanding tasks
  • TR roll-on/off attachment system for quick and secure disc changes
  • 10,000 RPM maximum operating speed
  • Closed coating design for consistent cutting action
  • Package quantity of 25
Catalog No. 50-277-2342 Supplier Grainger Supplier No. 13T248
These quick-change sanding discs are designed for effective material removal and surface preparation. Featuring a high-performance ceramic abrasive on a durable Y weight cloth backing, these discs provide reliable performance for heavy-duty applications. The TR roll-on/off attachment system allows for fast and secure disc changes, minimizing downtime during operation.
  • Ceramic abrasive material provides durability and cutting efficiency
  • 36 grit extra coarse abrasive grade for aggressive material removal
  • 4 in. diameter for versatile surface work
  • Durable Y weight cloth backing for demanding tasks
  • TR roll-on/off attachment system for quick and secure disc changes
  • 10,000 RPM maximum operating speed
  • Closed coating design for consistent cutting action
  • Package quantity of 25