Learn More
Medchemexpress LLC HY-135008 5mg Medchemexpress, J14 CAS:1043854-13-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1350085MG
Encompass_Preferred
Medchemexpress, HY-135008 5mg J14 CAS:1043854-13-2 J14 is a reversible sulfiredoxin inhibitor with an IC50 of 8.1 μM. J14 induces oxidative stress (intracellular ROS accumulation) by inhibiting sulfiredoxin, leading to cytotoxicity and cancer cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9856