Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ J14 Human SLP-76-Deficient Jurkat Cell Line

Catalog No. SCC190MI Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Cells:
≥ 1 x 106 Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Cells
SCC190MI ≥ 1 x 106 Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC190MI Supplier MilliporeSigma™ Supplier No. SCC190
Add to Cart
Add to Cart

J14 cells are a cell line used for elucidating cellular signaling mechanisms and interaction network biology.

    SLP-76 (SH2 domain–containing leukocyte protein of 76 kilodaltons), an adaptor protein and PTK substrate expressed in hematopoietic cells
  • SLP-76 and the scaffolding protein LAT act as a hub for various signaling effectors
  • Uses:
    • Elucidating cellular signals and interaction network biology
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in liquid nitrogen, Cell can be cultured for at least 10 passages after initial thawing without significantly affecting cell marker expression and functionality
    Cell Type Blood Cells
    For Use With (Application) Cell Culture
    No. of Cells ≥ 1 x 106 Cells/Vial

    For research purposes only

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.