Learn More
Medchemexpress LLC J147 | 1807913-16-1 | 99.5% | 350.34 | 25 MG
SDP
Catalog No. 5000352208
Encompass_Preferred
(E/Z)-J147 is a highly potent, orally active neuroprotective agent that enhances cognitive function. It can easily cross the blood-brain barrier (BBB). This compound inhibits monoamine oxidase B (MAO B) and the dopamine transporter. It shows promise for research related to Alzheimer's disease (AD).
  • Enhances cognitive function
  • Easily crosses the blood-brain barrier
  • Inhibits monoamine oxidase B (MAO B)
  • Inhibits the dopamine transporter
  • Promotes HT22 and primary cell survival in a dose-dependent manner
  • Enhances memory and dendritic spine number in old mice
Catalog No. 50-003-52208 Supplier Medchemexpress LLC Supplier No. 25MGHY13779
(E/Z)-J147 is a highly potent, orally active neuroprotective agent that enhances cognitive function. It can easily cross the blood-brain barrier (BBB). This compound inhibits monoamine oxidase B (MAO B) and the dopamine transporter. It shows promise for research related to Alzheimer's disease (AD).
  • Enhances cognitive function
  • Easily crosses the blood-brain barrier
  • Inhibits monoamine oxidase B (MAO B)
  • Inhibits the dopamine transporter
  • Promotes HT22 and primary cell survival in a dose-dependent manner
  • Enhances memory and dendritic spine number in old mice