Learn More
Medchemexpress LLC HY-126147 5mg Medchemexpress, J22352 CAS:2252395-44-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1261475MG
Encompass_Preferred
Medchemexpress, HY-126147 5mg J22352 CAS:2252395-44-9 J22352 is a PROTAC (proteolysis-targeting chimeras)-like and highly selective HDAC6 inhibitor with an IC50 value of 4.7 nM. J22352 promotes HDAC6 degradation and induces anticancer effects by inhibiting autophagy and eliciting the antitumor immune response in glioblastoma cancers, and leading to the restoration of host antitumor activity by reducing the immunosuppressive activity of PD-L1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9719