Learn More
ATCC J45.01; Acute T Cell Leukemia; Human (Homo sapiens)
Supplier:  ATCC CRL1990
Encompass
J45.01 is a T lymphoblast cell line that was isolated from the peripheral blood of a male patient with acute T cell leukemia. This cell line was deposited by GA Koretsky. It can be used in immune system disorders and immunology research.
Catalog No. 50-238-4568